missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorohydroquinone, tech. 90%, Thermo Scientific™

Product Code. 11424127
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424127 500 g 500g
11404127 25 g 25g
11414127 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424127 Supplier Thermo Scientific Alfa Aesar Supplier No. A19554.36
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-67-8
Molecular Formula C6H5ClO2
Molecular Weight (g/mol) 144.554
MDL Number MFCD00002341
InChI Key AJPXTSMULZANCB-UHFFFAOYSA-N
Synonym chlorohydroquinone, 2-chlorohydroquinone, monochlorohydroquinone, chloroquinol, 1,4-benzenediol, 2-chloro, 2-chloro-1,4-benzenediol, hydroquinone, chloro, 2-chloro-1,4-dihydroxybenzene, 2,5-dihydroxychlorobenzene, 1,4-dihydroxy-2-chlorobenzene
PubChem CID 301
ChEBI CHEBI:27675
IUPAC Name 2-chlorobenzene-1,4-diol
SMILES C1=CC(=C(C=C1O)Cl)O

Specifications

Melting Point 97°C to 102°C
Boiling Point 262°C to 263°C
Quantity 500 g
Formula Weight 144.56
Percent Purity 90%
Grade Technical
Chemical Name or Material 2-Chlorohydroquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.