missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenol, 99%

Product Code. p-7080676
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
This item is not returnable. View return policy

Product Code. 11474063

Brand: Thermo Scientific Alfa Aesar A10311.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-57-8 | C6H5ClO | 128.555 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-57-8
Molecular Formula C6H5ClO
Molecular Weight (g/mol) 128.555
MDL Number MFCD00002159
InChI Key ISPYQTSUDJAMAB-UHFFFAOYSA-N
Synonym o-chlorophenol, phenol, 2-chloro, 2-hydroxychlorobenzene, o-chlorphenol, o-chlorophenic acid, phenol, o-chloro, 2-chloro-1-hydroxybenzene, chlorophenol, septi-kleen, 2-chloro-phenol
PubChem CID 7245
ChEBI CHEBI:47083
IUPAC Name 2-chlorophenol
SMILES C1=CC=C(C(=C1)O)Cl

Specifications

Melting Point 7°C to 8°C
Density 1.26
Boiling Point 175°C to 176°C
Flash Point 63°C (145°F)
Odor Phenol-like
Refractive Index 1.559
Quantity 250 g
UN Number UN2021
Beilstein 1905114
Merck Index 14,2154
Formula Weight 128.56
Percent Purity 99%
Chemical Name or Material 2-Chlorophenol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.