missing translation for 'onlineSavingsMsg'
Learn More

(2-Chlorophenyl)diphenylmethanol, TRC

Product Code. 17983375
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17983375 100 mg 100mg
17902685 25 mg 25mg
17974025 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17983375 Supplier Toronto Research Chemicals Supplier No. TRCC377905100MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66774-02-5
Molecular Formula C19 H15 Cl O
Molecular Weight (g/mol) 294.77
Synonym (2-Chlorophenyl)diphenylmethanol, Clotrimazole Imp. A (EP), Benzenemethanol, 2-chloro-α,α-diphenyl-, Methanol, (o-chlorophenyl)diphenyl- (6CI), NSC 170882, TRAM 3, o-Chlorophenyldiphenylmethanol, Benzenemethanol, 2-chloro-α,α-diphenyl- (9CI, ACI), 2-Chloro-α,α-diphenylbenzenemethanol (ACI)
IUPAC Name (2-chlorophenyl)-diphenylmethanol
SMILES OC(c1ccccc1)(c2ccccc2)c3ccccc3Cl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C19H15ClO/c20-18-14-8-7-13-17(18)19(21,15-9-3-1-4-10-15)16-11-5-2-6-12-16/h1-14,21H
Formula Weight 294.0811
Percent Purity >95%
Chemical Name or Material (2-Chlorophenyl)diphenylmethanol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.