missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenyl isothiocyanate, 97%

Product Code. 11483767
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11483767 5 g 5g
11493767 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 11483767

Brand: Thermo Scientific Alfa Aesar A18238.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2740-81-0 | C7H4ClNS | 169.626 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2740-81-0
Molecular Formula C7H4ClNS
Molecular Weight (g/mol) 169.626
MDL Number MFCD00004801
InChI Key DASSPOJBUMBXLU-UHFFFAOYSA-N
Synonym 2-chlorophenyl isothiocyanate, 2-chlorophenylisothiocyanate, benzene, 1-chloro-2-isothiocyanato, benzene, chloroisothiocyanato, 1-chloro-2-isothiocyanato-benzene, isothiocyanic acid 2-chlorophenyl ester, 2-chlorobenzenisothiocyanate, acmc-1ccw5, o-chlorophenylisothiocyanate, o-chlorophenyl isothiocyanate
PubChem CID 123171
IUPAC Name 1-chloro-2-isothiocyanatobenzene
SMILES C1=CC=C(C(=C1)N=C=S)Cl

Specifications

Density 1.287
Boiling Point 260°C to 262°C
Quantity 5 g
Odor Pungent
Refractive Index 1.6608
UN Number UN2922
Beilstein 2081473
Formula Weight 169.64
Percent Purity 97%
Chemical Name or Material 2-Chlorophenyl isothiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.