missing translation for 'onlineSavingsMsg'
Learn More

2-chlorophenylacetyl Chloride, 97%

Product Code. 10266623 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10266623 5 g Glass bottle 5g
10298463 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10266623 Supplier Thermo Scientific Acros Supplier No. 395080050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51512-09-5 | C8H6Cl2O | 189.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51512-09-5
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.04
MDL Number MFCD00130223
InChI Key WIHSAOYVGKVRJX-UHFFFAOYSA-N
Synonym 2-chlorophenylacetyl chloride, 2-2-chlorophenyl acetyl chloride, acetyl chloride, 2-2-chlorophenyl, o-chlorophenylacetyl chloride, benzeneacetyl chloride, 2-chloro, 2-chlorophenylacetyl chloride stabilized with copper chip, acmc-1auqq, o-chlor-phenylacetylchlorid, o-chloro-phenylacetyl chloride, 2-chloro-phenylacetyl chloride
PubChem CID 182669
IUPAC Name 2-(2-chlorophenyl)acetyl chloride
SMILES ClC(=O)CC1=CC=CC=C1Cl

Specifications

CAS Min % 96.0
Color Yellow
CAS Max % 100.0
Density 1.3100g/mL
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5480 to 1.55
Assay Percent Range 97%
Specific Gravity 1.31
Linear Formula ClC6H4CH2COCl
Formula Weight 189.04
Percent Purity 97%
Chemical Name or Material 2-chlorophenylacetyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.