missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropyridine, 99%

Product Code. 11412657
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11412657 100 g 100g
11422657 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11412657 Supplier Thermo Scientific Alfa Aesar Supplier No. A14866.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 109-09-1 | C5H4ClN | 113.544 g/mol

2- Chloropyridine is used as an intermediate chemical in the production of various pyrithione, for use in cosmetics, personal care products and pharmaceutical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2- Chloropyridine is used as an intermediate chemical in the production of various pyrithione, for use in cosmetics, personal care products and pharmaceutical products.

Solubility
Soluble in water (27g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-09-1
Molecular Formula C5H4ClN
Molecular Weight (g/mol) 113.544
MDL Number MFCD00006228
InChI Key OKDGRDCXVWSXDC-UHFFFAOYSA-N
Synonym o-chloropyridine, pyridine, 2-chloro, alpha-chloropyridine, .alpha.-chloropyridine, unii-8hmd45ayej, 2-chloro-pyridine, ccris 1724, 8hmd45ayej, 6-chloro-pyridine, dsstox_cid_4810
PubChem CID 7977
ChEBI CHEBI:39174
IUPAC Name 2-chloropyridine
SMILES C1=CC=NC(=C1)Cl

Specifications

Melting Point -46°C
Density 1.208
pH 6 to 7
Boiling Point 167°C to 170°C
Flash Point 65°C (149°F)
Refractive Index 1.532
Quantity 100 g
UN Number UN2822
Beilstein 105788
Solubility Information Soluble in water (27g/L).
Formula Weight 113.55
Percent Purity 99%
Chemical Name or Material 2-Chloropyridine
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Chloropyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 2-Chloropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.