missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-3-fluoropyridine, 97+%, Thermo Scientific Chemicals

Product Code. 11484330
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484330 5 g 5g
11474330 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484330 Supplier Thermo Scientific Alfa Aesar Supplier No. H25767.06
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 97509-75-6 | Molecular Formula: C6H3FN2 | Molecular Weight: 122.102 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97509-75-6
Molecular Formula C6H3FN2
Molecular Weight (g/mol) 122.102
MDL Number MFCD06797501
InChI Key VZFPSCNTFBJZHB-UHFFFAOYSA-N
Synonym 2-cyano-3-fluoropyridine, 3-fluoropicolinonitrile, 3-fluoro-2-pyridinecarbonitrile, 3-fluoro-2-cyanopyridine, 3-fluoro-2-pyridinenitrile, 3-fluoro-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 3-fluoro, fluorocyanopyridine, pubchem16434, acmc-209s8i
PubChem CID 7060408
IUPAC Name 3-fluoropyridine-2-carbonitrile
SMILES C1=CC(=C(N=C1)C#N)F

Specifications

Melting Point 27°C to 30°C
Quantity 5 g
Formula Weight 122.1
Percent Purity ≥97%
Chemical Name or Material 2-Cyano-3-fluoropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.