missing translation for 'onlineSavingsMsg'
Learn More

2-Cyano-4'-methylbiphenyl, 98+%

Product Code. 11479167
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479167 25 g 25g
11469167 5 g 5g
11369629 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479167 Supplier Thermo Scientific Alfa Aesar Supplier No. B21273.14
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 114772-53-1 | C14H11N | 193.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114772-53-1
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.25
MDL Number MFCD00151805
InChI Key ZGQVZLSNEBEHFN-UHFFFAOYSA-N
Synonym 2-cyano-4'-methylbiphenyl, 4'-methyl-2-cyanobiphenyl, 4'-methylbiphenyl-2-carbonitrile, 2'-cyano-4-methylbiphenyl, 2-p-tolyl benzonitrile, 2-4-methylphenyl benzonitrile, 2-cyano-4'-methyl biphenyl, 4'-methyl-1,1'-biphenyl-2-carbonitrile, unii-o417ti15ta, 4'-methyl-2-cyano diphenyl
PubChem CID 145512
IUPAC Name 2-(4-methylphenyl)benzonitrile
SMILES CC1=CC=C(C=C1)C1=CC=CC=C1C#N

Specifications

Melting Point 49°C to 53°C
Density 1.17
Boiling Point >320°C
Quantity 25 g
UN Number UN3077
Beilstein 3605954
Formula Weight 193.26
Percent Purity ≥98%
Chemical Name or Material 2-Cyano-4'-methylbiphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.