missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanobenzoic acid, 94%

Product Code. 10163344 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10163344 1 g Glass bottle 1g
10123394 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10163344 Supplier Thermo Scientific Acros Supplier No. 398140010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3839-22-3 | C8H5NO2 | 147.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3839-22-3
Molecular Formula C8H5NO2
Molecular Weight (g/mol) 147.13
MDL Number MFCD00045796
InChI Key DTNSDCJFTHMDAK-UHFFFAOYSA-N
Synonym o-cyanobenzoic acid, cyanobenzoic acid, benzoic acid, 2-cyano, 2-carboxybenzonitrile, 2-carboxy-benzonitrile, 2-cyanobenzoicacid, 2-cyano-benzoic acid, pubchem12513, acmc-1bn2v
PubChem CID 138061
IUPAC Name 2-cyanobenzoic acid
SMILES C1=CC=C(C(=C1)C#N)C(=O)O

Specifications

CAS Min % 92.0
CAS Max % 100.0
Melting Point 212.0°C
Color Colorless to Yellow or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 92% min. (HPLC)
Packaging Glass bottle
Linear Formula NCC6H4CO2H
Quantity 1 g
Formula Weight 147.13
Percent Purity 94%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 2-Cyanobenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Cyanobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.