missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanobenzoic acid, 94%

Product Code. 11499350
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499350 1 g 1g
11409360 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499350 Supplier Thermo Scientific Alfa Aesar Supplier No. H50237.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3839-22-3 | C8H5NO2 | 147.133 g/mol

2-Cyanobenzoic acid has been used as reactant in the synthesis of T-box riboswitch antiterminator RNA-binding oxazolidinone derivatives, Benzoate modified β-cyclodextrin derivatives and Pyrrolo[1,2-f]triazines for use as JAK2 inhibitors. As a reactant involved in decarboxylation, decarboxylative halogenation and protodecarboxylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyanobenzoic acid has been used as reactant in the synthesis of T-box riboswitch antiterminator RNA-binding oxazolidinone derivatives, Benzoate modified β-cyclodextrin derivatives and Pyrrolo[1,2-f]triazines for use as JAK2 inhibitors. As a reactant involved in decarboxylation, decarboxylative halogenation and protodecarboxylation.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3839-22-3
Molecular Formula C8H5NO2
Molecular Weight (g/mol) 147.133
MDL Number MFCD00045796
InChI Key DTNSDCJFTHMDAK-UHFFFAOYSA-N
Synonym o-cyanobenzoic acid, cyanobenzoic acid, benzoic acid, 2-cyano, 2-carboxybenzonitrile, 2-carboxy-benzonitrile, 2-cyanobenzoicacid, 2-cyano-benzoic acid, pubchem12513, acmc-1bn2v
PubChem CID 138061
IUPAC Name 2-cyanobenzoic acid
SMILES C1=CC=C(C(=C1)C#N)C(=O)O

Specifications

Melting Point 210°C to 212°C
Quantity 1 g
Formula Weight 147.13
Percent Purity 96%
Chemical Name or Material 2-Cyanobenzoic acid
compliance-icons
Product Identifier
  • 2-Cyanobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Cyanobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.