missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclopentylphenol, 98+%

Product Code. 10623215 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10623215 1 g Glass bottle 1g
10516104 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10623215 Supplier Thermo Scientific Acros Supplier No. 426510010
 Restricted Item
Estimated Shipment: 19-05-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1518-84-9 | C11H14O | 162.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1518-84-9
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00009952
InChI Key JHEKSKQMOBLXQS-UHFFFAOYSA-N
Synonym phenol, 2-cyclopentyl, 2-cyclopentyl-phenol, o-cyclopentylphenol, o-cyclopentyl-phenol, 2-cyclopentyl phenol, acmc-20anxa, phenol, o-cyclopentyl, 2-cyclopentylphenol, o-cyclopentyl, 2-cyclopentylphenol, technical grade
PubChem CID 80285
IUPAC Name 2-cyclopentylphenol
SMILES OC1=CC=CC=C1C1CCCC1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 36°C
Color Colorless to Yellow
Boiling Point 148°C to 150°C (18.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 162.23
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2-Cyclopentylphenol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Cyclopentylphenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.