missing translation for 'onlineSavingsMsg'
Learn More

2-Decanone, 97%

Product Code. 11359629
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359629 5 g 5g
11408126 25 g 25g
11418126 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11359629 Supplier Thermo Scientific Alfa Aesar Supplier No. A14145.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 693-54-9 | C10H20O | 156.269 g/mol

2-Decanone was used in the synthesis of brushed block copolymers via conjugation through an acid-labile hydrazone linker to poly(ethylene glycol)-poly(aspartate hydrazide) block copolymers; methyl ketones. 2-decanone used as a solvent and an intermediate in the synthesis of pharmaceuticals and agrochemicals. It is also used in perfumes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Decanone was used in the synthesis of brushed block copolymers via conjugation through an acid-labile hydrazone linker to poly(ethylene glycol)-poly(aspartate hydrazide) block copolymers; methyl ketones. 2-decanone used as a solvent and an intermediate in the synthesis of pharmaceuticals and agrochemicals. It is also used in perfumes.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-54-9
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00009571
InChI Key ZAJNGDIORYACQU-UHFFFAOYSA-N
Synonym 2-decanone, methyl octyl ketone, methyl n-octyl ketone, octyl methyl ketone, unii-gx543olt0r, n-c8h17coch3, gx543olt0r, chembl47127
PubChem CID 12741
ChEBI CHEBI:77929
IUPAC Name decan-2-one
SMILES CCCCCCCCC(=O)C

Specifications

Melting Point 3°C
Density 0.824
Boiling Point 210°C to 212°C
Flash Point 86°C (187°F)
Refractive Index 1.425
Quantity 5 g
Beilstein 1747463
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 156.27
Percent Purity 97%
Chemical Name or Material 2-Decanone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.