missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Deoxyadenosine, 99%

Product Code. 15486039
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
100 g
Unit Size:
100g
25g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15486039 5 g 5g
15496039 25 g 25g
15406049 50 g 50g
15416049 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15486039 Supplier Thermo Scientific Chemicals Supplier No. J63886.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 958-09-8 | C10H13N5O3 | 251.25 g/mol

2'-Deoxyadenosine is used in paradoxical stimulation study of human sperm motility. It plays an essential role as sperm motility stimulants used in sperm micro-injection on mouse and human embryo development. It finds an application in some cells as an energy source under energy stress conditions. It is involved in various biological processes to compare the functions of adenosine analogues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Deoxyadenosine is used in paradoxical stimulation study of human sperm motility. It plays an essential role as sperm motility stimulants used in sperm micro-injection on mouse and human embryo development. It finds an application in some cells as an energy source under energy stress conditions. It is involved in various biological processes to compare the functions of adenosine analogues.

Solubility
Soluble in water and caustic soda.

Notes
Air sensitive. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 958-09-8
Molecular Formula C10H13N5O3
Molecular Weight (g/mol) 251.25
MDL Number MFCD00005754
InChI Key OLXZPDWKRNYJJZ-RRKCRQDMSA-N
Synonym 2'-deoxyadenosine, deoxyadenosine, 2-deoxyadenosine, 2r,3s,5r-5-6-amino-9h-purin-9-yl-2-hydroxymethyl tetrahydrofuran-3-ol, adenyldeoxyriboside, adenine deoxyribonucleoside, adenine deoxyribose, desoxyadenosine, ccris 1782, unii-p582c98ulc
PubChem CID 13730
ChEBI CHEBI:17256
IUPAC Name (2R,3S,5R)-5-(6-aminopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
SMILES NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1

Specifications

Color White
Quantity 5 g
Solubility Information Soluble in water and caustic soda.
Optical Rotation −25° (c=0.5 in Water)
Formula Weight 251.24
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2'-Deoxyadenosine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.