missing translation for 'onlineSavingsMsg'
Learn More

2-(Difluoromethoxy)-4-fluorobenzyl bromide, 97%, Thermo Scientific™

Product Code. 15418165
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15418165

Brand: Thermo Scientific Alfa Aesar H32676.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1017779-42-8
Molecular Formula C8H6BrF3O
Molecular Weight (g/mol) 255.034
MDL Number MFCD09832363
InChI Key IGODPIYHAUGSCK-UHFFFAOYSA-N
Synonym 2-difluoromethoxy-4-fluorobenzyl bromide, 1-bromomethyl-2-difluoromethoxy-4-fluorobenzene, 2-bromomethyl-5-fluorophenoxy difluoromethane
PubChem CID 46737679
IUPAC Name 1-(bromomethyl)-2-(difluoromethoxy)-4-fluorobenzene
SMILES C1=CC(=C(C=C1F)OC(F)F)CBr

Specifications

Refractive Index 1.5035
Quantity 250 mg
UN Number UN3265
Formula Weight 255.03
Percent Purity 97%
Chemical Name or Material 2-(Difluoromethoxy)-4-fluorobenzyl bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.