missing translation for 'onlineSavingsMsg'
Learn More

2-(Difluoromethoxy)aniline, 97%, Thermo Scientific™

Product Code. p-7024975
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11375299

Brand: Thermo Scientific Alfa Aesar B20255.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22236-04-0
Molecular Formula C7H7F2NO
Molecular Weight (g/mol) 159.14
MDL Number MFCD00190104
InChI Key CGNAIUUCOVWLLL-UHFFFAOYSA-N
Synonym 2-difluoromethoxy aniline, o-difluoromethoxyaniline, 2-difluoromethoxy-phenylamine, benzenamine, 2-difluoromethoxy, 2-difluoromethoxy phenylamine, pubchem8465, acmc-1cll9, akos bb184, asischem y89474, ksc494k1l
PubChem CID 934620
IUPAC Name 2-(difluoromethoxy)aniline
SMILES NC1=CC=CC=C1OC(F)F

Specifications

Density 1.265
Boiling Point 205°C
Flash Point 92°C (197°F)
Odor Characteristic
Refractive Index 1.504
Quantity 1 g
UN Number UN2810
Beilstein 2718520
Formula Weight 159.14
Percent Purity 97%
Chemical Name or Material 2-(Difluoromethoxy)aniline
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.