missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Difluoromethyl)-1,4-difluorobenzene, 97%, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Boiling Point | 120°C |
| Flash Point | 28°C |
| Assay Percent Range | 96% min. (GC) |
| Packaging | Glass Bottle |
| Quantity | 1 g |
| Formula Weight | 164.1 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-(Difluoromethyl)-1, 4-difluorobenzene |
Safety and Handling
Product Identifier
- 2-(Difluoromethyl)-1,4-difluorobenzene, 97%
- Warning
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido