missing translation for 'onlineSavingsMsg'
Learn More

2-(Difluoromethyl)-1,4-difluorobenzene, 97%, Thermo Scientific™

Product Code. 15269756 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15269756

Brand: Thermo Scientific Acros 451370050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 195886-79-4
Molecular Formula C7H4F4
Molecular Weight (g/mol) 164.1
InChI Key FDUVCOVVYFCQEH-UHFFFAOYSA-N
Synonym 2-difluoromethyl-1,4-difluorobenzene, 2,5-difluorobenzal fluoride, 1-difluoromethyl-2,5-difluorobenzene, 2,4-difluoro-1-difluoromethyl benzene
PubChem CID 46311703
IUPAC Name 2-(difluoromethyl)-1,4-difluorobenzene
SMILES C1=CC(=C(C=C1F)C(F)F)F

Specifications

Boiling Point 120°C
Flash Point 28°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 164.1
Percent Purity 97%
Chemical Name or Material 2-(Difluoromethyl)-1, 4-difluorobenzene
compliance-icons compliance-icons
Product Identifier
  • 2-(Difluoromethyl)-1,4-difluorobenzene, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.