missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyphenol, 98%, Thermo Scientific Chemicals

Product Code. 11436466
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11436466 500 g 500g
11416466 25 g 25g
11426466 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11436466 Supplier Thermo Scientific Alfa Aesar Supplier No. B24755.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 94-71-3 | Molecular Formula: C8H10O2 | Molecular Weight: 138.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-71-3
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00002187
InChI Key MOEFFSWKSMRFRQ-UHFFFAOYSA-N
Synonym o-ethoxyphenol, phenol, 2-ethoxy, guaethol, guethol, 2-ethyloxyphenol, guaiethol, pyrocatechol monoethyl ether, catechol monoethyl ether, phenol, o-ethoxy, unii-878iw8p9pw
PubChem CID 66755
IUPAC Name 2-ethoxyphenol
SMILES CCOC1=CC=CC=C1O

Specifications

Melting Point 24°C to 27°C
Density 1.1
Boiling Point 214°C to 216°C
Flash Point 86°C (187°F)
Odor Phenol-like
Quantity 500 g
Formula Weight 138.17
Percent Purity 98%
Chemical Name or Material 2-Ethoxyphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.