missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyphenol, 98%, Thermo Scientific Chemicals

Product Code. 10763413 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10763413 500 g Glass bottle 500g
10151192 5 g Glass bottle 5g
10571882 100 g Glass bottle 100g
10539213 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10763413 Supplier Thermo Scientific Acros Supplier No. 370035000
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 94-71-3 | Molecular Formula: C8H10O2 | Molecular Weight: 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-71-3
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00002187
InChI Key MOEFFSWKSMRFRQ-UHFFFAOYSA-N
Synonym o-ethoxyphenol, phenol, 2-ethoxy, guaethol, guethol, 2-ethyloxyphenol, guaiethol, pyrocatechol monoethyl ether, catechol monoethyl ether, phenol, o-ethoxy, unii-878iw8p9pw
PubChem CID 66755
IUPAC Name 2-ethoxyphenol
SMILES CCOC1=CC=CC=C1O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 20.0°C to 25.0°C
Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 216.0°C to 217.0°C
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5OC6H4OH
Refractive Index 1.5280 to 1.5300
Quantity 500 g
Specific Gravity 1.09
Formula Weight 138.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Ethoxyphenol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Ethoxyphenol
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H412-Harmful to aquatic life with long lasting effects.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.