missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-6-methylphenol, 97%

Product Code. 11460708
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460708 1 g 1g
11470708 5 g 5g
11480708 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460708 Supplier Thermo Scientific Alfa Aesar Supplier No. B25213.03
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1687-64-5 | C9H12O | 136.194 g/mol

2-Ethyl-6-methylphenol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethyl-6-methylphenol is used as pharmaceutical intermediate.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1687-64-5
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00962292
InChI Key CIRRFAQIWQFQSS-UHFFFAOYSA-N
PubChem CID 519333
IUPAC Name 2-ethyl-6-methylphenol
SMILES CCC1=CC=CC(=C1O)C

Specifications

Refractive Index 1.531
Quantity 1 g
Synonym phenol, 2-ethyl-6-methyl, o-cresol, 6-ethyl, 6-ethyl-o-cresol, unii-359wuc62ri, 2-ethyl-6-methyl-phenol, 6-ethyl-2-methylphenol, acmc-20anwj, 2-methyl-6-ethylphenol, 2-ethyl-6-methyl phenol, phenol,2-ethyl-6-methyl
Formula Weight 136.19
Percent Purity 97%
Chemical Name or Material 2-Ethyl-6-methylphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.