missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(Ethylthio)benzeneboronic acid, 97%

Product Code. 15400098
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15400098 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15400098 Supplier Alfa Aesar™ Supplier No. H53350.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 362045-33-8
Molecular Formula C8H11BO2S
Molecular Weight (g/mol) 182.04
MDL Number MFCD03095256
InChI Key SCGSNVLNGRYSAA-UHFFFAOYSA-N
Synonym 2-ethylthiophenylboronic acid, 2-ethylthio phenylboronic acid, 2-ethylthiophenyl boronic acid, 2-ethylthio benzeneboronic acid, 2-ethylsulfanyl phenylboronic acid, boronic acid, b-2-ethylthio phenyl, 2-ethylsulfanyl phenyl boronic acid, boronic acid, 2-ethylthio phenyl, 2-ethylsulfanylphenyl boronic acid, pubchem1843
PubChem CID 3785655
SMILES CCSC1=CC=CC=C1B(O)O

Specifications

Melting Point 42°C to 46°C
Quantity 1 g
IUPAC Name [2-(ethylsulfanyl)phenyl]boronic acid
Formula Weight 182.04
Percent Purity 97%
Chemical Name or Material 2-(Ethylthio)benzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.