missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylthiophenol, 95%

Product Code. 11312457
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312457 10 g 10g
11302457 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312457 Supplier Thermo Scientific Alfa Aesar Supplier No. L11498.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4500-58-7 | C8H10S | 138.228 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4500-58-7
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.228
MDL Number MFCD00010022
InChI Key ABROBCBIIWHVNS-UHFFFAOYSA-N
Synonym 2-ethylthiophenol, benzenethiol, 2-ethyl, unii-62gbu45i5p, 2-ethyl thiophenol, o-ethylbenzenethiol, fema no. 3345, ethylbenzenethiol, o, 2-ethylphenylmercaptan, benzenethiol, o-ethyl, o-ethylthiophenol
PubChem CID 3734338
IUPAC Name 2-ethylbenzenethiol
SMILES CCC1=CC=CC=C1S

Specifications

Density 1.025
Boiling Point 203°C to 205°C
Flash Point 80°C (176°F)
Odor Stench
Refractive Index 1.569
Quantity 10 g
UN Number UN2810
Beilstein 2553288
Sensitivity Air Sensitive
Formula Weight 138.23
Percent Purity 95%
Chemical Name or Material 2-Ethylthiophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.