missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-nitrobenzoic acid, 97%

Product Code. 15459828
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15459828 5 g 5g
15449828 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15459828 Supplier Thermo Scientific Alfa Aesar Supplier No. H63235.06
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 317-46-4 | C7H4FNO4 | 185.11 g/mol

2-Fluoro-3-nitrobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-3-nitrobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 317-46-4
Molecular Formula C7H4FNO4
Molecular Weight (g/mol) 185.11
MDL Number MFCD06797825
InChI Key WLGUSLGYTNJJFV-UHFFFAOYSA-N
Synonym 2-fluoro-3-nitrobenzoic acid, 2-fluoro-3-nitrobenzoicacid, 2-fluoro-3-nitro benzoic acid, 3-carboxy-2-fluoronitrobenzene, pubchem1398, acmc-1ajf3, ksc494k8n, benzoic acid,2-fluoro-3-nitro
PubChem CID 18672992
IUPAC Name 2-fluoro-3-nitrobenzoic acid
SMILES OC(=O)C1=C(F)C(=CC=C1)[N+]([O-])=O

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 185.11
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Fluoro-3-nitrobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.