missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-nitrotoluene, 98%

Product Code. 15489718
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15489718 100 g 100g
15479718 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15489718 Supplier Thermo Scientific Alfa Aesar Supplier No. H63173.22
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 437-86-5 | C7H6FNO2 | 155.128 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 437-86-5
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD03412242
InChI Key NBCNUIXYBLFJMI-UHFFFAOYSA-N
Synonym 2-fluoro-3-nitrotoluene, 2-fluoro-3-methylnitrobenzene, benzene, 2-fluoro-1-methyl-3-nitro, pubchem1591, 2-fluor-3-nitrotoluol, acmc-1agoj, ksc235o7n, 2-fluoro-3-methyl-nitrobenzene, 2-fluoro-1-methyl-3-nitro-benzene, 1-methyl-2-fluoro-3-nitro-benzene
PubChem CID 13470957
IUPAC Name 2-fluoro-1-methyl-3-nitrobenzene
SMILES CC1=C(C(=CC=C1)[N+](=O)[O-])F

Specifications

Melting Point 19°C (decomposition)
Color Colorless to Yellow
Density 1.276 g/mL
Boiling Point 111°C (12 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.529
Quantity 100 g
UN Number UN2810
Beilstein 2442082
Formula Weight 155.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Fluoro-3-nitrotoluene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.