missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4,6-bis(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™

Product Code. 15464687
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15464687 1 g 1g
15474687 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15464687 Supplier Thermo Scientific Alfa Aesar Supplier No. H32020.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 1017777-37-5
Molecular Formula C9H3F7O
Molecular Weight (g/mol) 260.11
MDL Number MFCD09258668
InChI Key CXWRFYUFIINRJQ-UHFFFAOYSA-N
Synonym 2-fluoro-4,6-bis trifluoromethyl benzaldehyde, 2,4-bis trifluoromethyl-6-fluorobenzaldehyde, benzaldehyde,2-fluoro-4,6-bis trifluoromethyl
PubChem CID 46737497
IUPAC Name 2-fluoro-4,6-bis(trifluoromethyl)benzaldehyde
SMILES FC1=CC(=CC(=C1C=O)C(F)(F)F)C(F)(F)F

Spécification

Refractive Index 1.4154
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 260.11
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4,6-bis(trifluoromethyl)benzaldehyde
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.