missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-hydroxypyridine, 97%

Product Code. 15424317
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15424317 1 g 1g
15434317 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15424317 Supplier Thermo Scientific Alfa Aesar Supplier No. H31697.03
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

Denne vare kan ikke returneres. Se returpolicy

CAS: 22282-69-5 | C5H4FNO | 113.091 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 22282-69-5
Molecular Formula C5H4FNO
Molecular Weight (g/mol) 113.091
MDL Number MFCD04114159
InChI Key BRLSDLHMGRDAPF-UHFFFAOYSA-N
Synonym 2-fluoropyridin-4-ol, 2-fluoro-4-hydroxypyridine, 2-fluoro-pyridin-4-ol, 4-pyridinol, 2-fluoro, 2-fluoro-4-pyridinol, 4 1h-pyridinone,2-fluoro, 2-fluoranyl-1h-pyridin-4-one, 4 1h-pyridinone, 2-fluoro, 4 1h-pyridinone, 2-fluoro-9ci
PubChem CID 2762834
IUPAC Name 2-fluoro-1H-pyridin-4-one
SMILES C1=CNC(=CC1=O)F

Tekniske data

Melting Point 146°C to 149°C
Quantity 1 g
Formula Weight 113.09
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-hydroxypyridine

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.