missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-methoxycarbonylphenylboronic acid, 98%, Thermo Scientific™

Product Code. 15505176 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15505176 5 g 5g
15595166 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15505176 Supplier Thermo Scientific Acros Supplier No. 455870050
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 603122-84-5
Molecular Formula C8H8BFO4
Molecular Weight (g/mol) 197.96
MDL Number MFCD08436034
InChI Key BKWRLCIYMAYFPA-UHFFFAOYSA-N
Synonym 2-fluoro-4-methoxycarbonyl phenylboronic acid, 2-fluoro-4-methoxycarbonyl benzeneboronic acid, 4-methoxycarbonyl-2-fluorophenylboronic acid, 2-fluoro-4-methoxycarbonyl phenyl boronic acid, o-fluoro p-methoxy carbonyl phenyl boronic acid, pubchem17434, acmc-209mj4, 2-fluoro-4-methoxycarbonyl phenylboronicacid, 2-fluoro-4-methoxycarbonyl-phenylboronic acid
PubChem CID 24824345
IUPAC Name (2-fluoro-4-methoxycarbonylphenyl)boronic acid
SMILES COC(=O)C1=CC(F)=C(C=C1)B(O)O

Specifications

Melting Point 127.0°C to 129.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 197.96
Percent Purity 98%
Chemical Name or Material 2-Fluoro-4-methoxycarbonylphenylboronic acid
compliance-icons
Product Identifier
  • 2-Fluoro-4-methoxycarbonylphenylboronic acid, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.