missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-methylaniline, 99%, Thermo Scientific™

Product Code. 10776043 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10776043 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10776043 Supplier Thermo Scientific Acros Supplier No. 320040010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 452-80-2
CAS Min % 98.5
CAS Max % 100.0
Density 1.1080g/mL
Boiling Point 70.0°C to 71.0°C (7.0mmHg)
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C7H8FN
Linear Formula FC6H3(CH3)NH2
Refractive Index 1.5320 to 1.5340
MDL Number MFCD00040975
Quantity 1g
Synonym 2-fluoro-4-methyaniline, 2-fluoro-p-toluidine, 4-amino-3-fluorotoluene, p-toluidine, 2-fluoro, benzenamine, 2-fluoro-4-methyl, 2-fluoro-4-methyl aniline, 2-fluoro-4-methyl-aniline, 2-fluoro-4-methyl-phenylamine, 2-fluoro-4-methylphenylamine, benzenamine, 2-fluoro-4-methyl-9ci
InChI Key ZQEXBVHABAJPHJ-UHFFFAOYSA-N
SMILES CC1=CC=C(N)C(F)=C1
IUPAC Name 2-fluoro-4-methylaniline
Molecular Weight (g/mol) 125.15
PubChem CID 67984
Formula Weight 125.16
Percent Purity 99%
Chemical Name or Material 2-Fluoro-4-methylaniline, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.