missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-methylaniline, 99%

Product Code. 11477927
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11477927 25 g 25g
11467927 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11477927 Supplier Thermo Scientific Alfa Aesar Supplier No. A16591.14
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 452-80-2 | C7H8FN | 125.15 g/mol

2-Fluoro-4-methylaniline is used in the preparation of 6-chloro-5-fluoroindole via Leimgruber-Batcho reaction. It is also used in the preparation of an (S)-amino alcohol, 2-amino-3-(2-fluoro-4-methylphenyl)-propan-1-ol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-4-methylaniline is used in the preparation of 6-chloro-5-fluoroindole via Leimgruber-Batcho reaction. It is also used in the preparation of an (S)-amino alcohol, 2-amino-3-(2-fluoro-4-methylphenyl)-propan-1-ol.

Solubility
Not miscible in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452-80-2
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.15
MDL Number MFCD00040975
InChI Key ZQEXBVHABAJPHJ-UHFFFAOYSA-N
Synonym 2-fluoro-4-methyaniline, 2-fluoro-p-toluidine, 4-amino-3-fluorotoluene, p-toluidine, 2-fluoro, benzenamine, 2-fluoro-4-methyl, 2-fluoro-4-methyl aniline, 2-fluoro-4-methyl-aniline, 2-fluoro-4-methyl-phenylamine, 2-fluoro-4-methylphenylamine, benzenamine, 2-fluoro-4-methyl-9ci
PubChem CID 67984
IUPAC Name 2-fluoro-4-methylaniline
SMILES CC1=CC=C(N)C(F)=C1

Specifications

Density 1.108
Boiling Point 70°C to 71°C (7 mmHg)
Flash Point 79°C (174°F)
Refractive Index 1.533
Quantity 25 g
UN Number UN2810
Beilstein 2637578
Solubility Information Not miscible in water.
Formula Weight 125.15
Percent Purity 99%
Chemical Name or Material 2-Fluoro-4-methylaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.