missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-methylbenzaldehyde, 97%

Product Code. 11382499
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11382499 1 g 1g
11392499 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382499 Supplier Thermo Scientific Alfa Aesar Supplier No. H26626.03
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 146137-80-6 | C8H7FO | 138.141 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146137-80-6
Molecular Formula C8H7FO
Molecular Weight (g/mol) 138.141
MDL Number MFCD03094325
InChI Key MVDRIMBGRZBWPE-UHFFFAOYSA-N
Synonym 2-fluoro-4-methyl-benzaldehyde, 2-fluoro-4-methyl benzaldehyde, benzaldehyde, 2-fluoro-4-methyl, 2-fluoro-p-tolualdehyde, pubchem1451, intermediates-zcf02120, acmc-1c24x, ksc494i0d, benzaldehyde, 2-fluoro-4-methyl-9ci
PubChem CID 2778471
IUPAC Name 2-fluoro-4-methylbenzaldehyde
SMILES CC1=CC(=C(C=C1)C=O)F

Specifications

Refractive Index 1.524
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 138.14
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-methylbenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.