missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-nitrobenzyl alcohol, 97%, Thermo Scientific Chemicals

Product Code. 15436307
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15436307 1 g 1g
15446307 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15436307 Supplier Thermo Scientific Alfa Aesar Supplier No. H33494.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 660432-43-9
Molecular Formula C7H6FNO3
Molecular Weight (g/mol) 171.13
MDL Number MFCD11110168
InChI Key XYGZVFTWIGGORD-UHFFFAOYSA-N
Synonym 2-fluoro-4-nitrophenyl methanol, 2-fluoro-4-nitrobenzyl alcohol, benzenemethanol, 2-fluoro-4-nitro, 2-fluoro-4-nitrobenzylalcohol, 2-fluoro-4-nitro-phenyl-methanol, 3-fluoro-4-hydroxymethyl nitrobenzene
PubChem CID 17981733
IUPAC Name (2-fluoro-4-nitrophenyl)methanol
SMILES OCC1=C(F)C=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 103°C to 105°C
Color White to Yellow
Quantity 1 g
Formula Weight 171.1
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Fluoro-4-nitrobenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.