missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-nitrotoluene, 98%, Thermo Scientific™

Product Code. 10696782 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10696782 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10696782 Supplier Thermo Scientific Acros Supplier No. 207090050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1427-07-2
CAS Min % 97.5
CAS Max % 100.0
Boiling Point 65.0°C to 68.0°C (2.0mmHg)
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H6FNO2
Linear Formula CH3C6H3(NO2)F
MDL Number MFCD00007199
Quantity 5g
Beilstein 05, III, 743
Synonym 2-fluoro-4-nitrotoluene, toluene, 2-fluoro-4-nitro, benzene, 2-fluoro-1-methyl-4-nitro, 3-fluoro-4-methylnitrobenzene, 1-methyl-2-fluoro-4-nitrobenzene, 2-fluoro-1-methyl-4-nitro-benzene, pubchem1592, 2-fluoro4-nitrotoluene, acmc-209cov, 2-fluor-4-nitro-toluol
InChI Key WIQISTBTOQNVCE-UHFFFAOYSA-N
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])F
IUPAC Name 2-fluoro-1-methyl-4-nitrobenzene
Molecular Weight (g/mol) 155.13
PubChem CID 74025
Formula Weight 155.13
Percent Purity 98%
Chemical Name or Material 2-Fluoro-4-nitrotoluene, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluoro-4-nitrotoluene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.