missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(trifluoromethoxy)benzoic acid, 97%, Thermo Scientific Chemicals

Product Code. 15406607
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15406607 1 g 1g
15416607 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15406607 Supplier Thermo Scientific Alfa Aesar Supplier No. H33749.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

2-Fluoro-4-(trifluoromethyl)benzoic acid may be used in pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-4-(trifluoromethyl)benzoic acid may be used in pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073477-22-1
Molecular Formula C8H4F4O3
Molecular Weight (g/mol) 224.11
MDL Number MFCD16652439
InChI Key TUXSSQNLBPWREW-UHFFFAOYSA-N
Synonym 2-fluoro-4-trifluoromethoxy benzoic acid
PubChem CID 58072303
IUPAC Name 2-fluoro-4-(trifluoromethoxy)benzoic acid
SMILES OC(=O)C1=C(F)C=C(OC(F)(F)F)C=C1

Specifications

Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 224.11
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-(trifluoromethoxy)benzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.