missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(trifluoromethyl)benzaldehyde, 97%

Product Code. 11385189
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11385189 25 g 25g
11365189 1 g 1g
11375189 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11385189 Supplier Thermo Scientific Alfa Aesar Supplier No. A18500.14
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 89763-93-9 | C8H4F4O | 192.113 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89763-93-9
Molecular Formula C8H4F4O
Molecular Weight (g/mol) 192.113
MDL Number MFCD00061310
InChI Key KFEHNXLFIGPWNB-UHFFFAOYSA-N
Synonym 2-fluoro-4-trifluoromethyl benzaldehyde, benzaldehyde, 2-fluoro-4-trifluoromethyl, pubchem4235, acmc-209r2s, ksc496e9p, attercop-chm at111109, a,a,a,2-tetrafluoro-p-tolualdehyde, 2-fluoro-4-trifluoromethyl benzaldehy, 2-fluoro-4-trifluoromethyl-benzaldehyde, 2-fluoro4-trifluoromethyl benzaldehyde
PubChem CID 522929
IUPAC Name 2-fluoro-4-(trifluoromethyl)benzaldehyde
SMILES C1=CC(=C(C=C1C(F)(F)F)F)C=O

Specifications

Density 1.41
Boiling Point 118°C to 119°C
Flash Point 110° (230°F)
Refractive Index 1.45
Quantity 25 g
Beilstein 7703632
Sensitivity Air Sensitive
Formula Weight 192.11
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)benzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.