missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific™

Product Code. 11428817
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11428817

Brand: Thermo Scientific Alfa Aesar A19950.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126917-10-0
Molecular Formula C8H3ClF4O
Molecular Weight (g/mol) 226.555
MDL Number MFCD00061155
InChI Key OOAHPLWBUUTFMV-UHFFFAOYSA-N
PubChem CID 145580
IUPAC Name 2-fluoro-4-(trifluoromethyl)benzoyl chloride
SMILES C1=CC(=C(C=C1C(F)(F)F)F)C(=O)Cl

Specifications

Density 1.5
Boiling Point 188°C to 189°C
Flash Point 81°C (177°F)
Refractive Index 1.465
Quantity 5 g
UN Number UN3265
Beilstein 4251456
Sensitivity Moisture sensitive
Synonym 2-fluoro-4-trifluoromethyl benzoyl chloride, 2-fluoro-4-trifluoromethyl benzoylchloride, benzoyl chloride, 2-fluoro-4-trifluoromethyl, 2-fluoro-4-trifluoromethyl-benzoyl chloride, benzoyl chloride,2-fluoro-4-trifluoromethyl, 2-fluoro-4-trifluoromethyl-1-benzenecarbonyl chloride, alpha,alpha,alpha,2-tetrafluoro-p-toluoyl chloride, acmc-1c5cr, 2-fluoro-4-trifluoromethyl-benzoylchloride, 4-trifluoromethyl-2-fluorobenzoyl chloride
Formula Weight 226.56
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)benzoyl chloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.