missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-5-(isopropylcarbamoyl)benzeneboronic acid, 98%

Product Code. 15499935
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499935 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15499935 Supplier Alfa Aesar™ Supplier No. H52605.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 874289-49-3
Molecular Formula C10H13BFNO3
Molecular Weight (g/mol) 225.03
MDL Number MFCD08235103
InChI Key NCRUGKRIPMSQHI-UHFFFAOYSA-N
Synonym 2-fluoro-5-isopropylcarbamoyl phenyl boronic acid, n-isopropyl 3-borono-4-fluorobenzamide, 2-fluoro-5-isopropylcarbamoyl benzeneboronic acid, 2-fluoro-5-isopropylcarbamoyl phenylboronic acid, acmc-209qmq, 3-borono-4-fluoro-n-isopropylbenzamide, 3-borono-4-fluoro-n-propan-2-yl benzamide, 2-fluoro-5-isopropylcarbamoyl phenyl boronicacid, 2-fluoro-5-isopropylcarbamoyl-phenyl boronic acid, 2-fluoranyl-5-propan-2-ylcarbamoyl phenyl boronic acid
PubChem CID 44717605
IUPAC Name [2-fluoro-5-(propan-2-ylcarbamoyl)phenyl]boronic acid
SMILES CC(C)NC(=O)C1=CC(B(O)O)=C(F)C=C1

Specifications

Melting Point 130°C to 134°C
Quantity 250 mg
Formula Weight 225.03
Percent Purity 98%
Chemical Name or Material 2-Fluoro-5-(isopropylcarbamoyl)benzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.