missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methoxy-3-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™

Product Code. 15498415
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15498415 5 g 5g
15488415 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15498415

Brand: Thermo Scientific Alfa Aesar H33361.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1373920-90-1
Molecular Formula C9H6F4O3
Molecular Weight (g/mol) 238.138
MDL Number MFCD22201034
InChI Key OVJKRERPBULYBW-UHFFFAOYSA-N
Synonym 2-fluoro-5-methoxy-3-trifluoromethyl benzoic acid
PubChem CID 86277636
IUPAC Name 2-fluoro-5-methoxy-3-(trifluoromethyl)benzoic acid
SMILES COC1=CC(=C(C(=C1)C(=O)O)F)C(F)(F)F

Specifications

Quantity 5 g
Formula Weight 238.14
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-methoxy-3-(trifluoromethyl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.