missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methylbenzonitrile, 99%

Product Code. p-7028759
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11316549

Brand: Thermo Scientific Alfa Aesar H26166.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 64113-84-4 | C8H6FN | 135.141 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64113-84-4
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.141
MDL Number MFCD00134542
InChI Key CMAOLVNGLTWICC-UHFFFAOYSA-N
Synonym 6-fluoro-m-tolunitrile, benzonitrile, 2-fluoro-5-methyl, 2-fluoro-5-methyl-benzonitrile, 3-cyano-4-fluorotoluene, 2-fluoro-5-methylbenzenecarbonitrile, pubchem1552, acmc-209nk8, ksc352m4h, 2-fluoro-5 methyl benzonitrile, 2-fluoro-5-methyl benzonitrile
PubChem CID 2774584
IUPAC Name 2-fluoro-5-methylbenzonitrile
SMILES CC1=CC(=C(C=C1)F)C#N

Specifications

Melting Point 49°C to 52°C
Flash Point 94°C (201°F)
Quantity 1 g
UN Number UN3439
Formula Weight 135.14
Percent Purity 99%
Chemical Name or Material 2-Fluoro-5-methylbenzonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.