missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-nitrobenzoic acid, 98%, Thermo Scientific™

Product Code. 10051034 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10051034 1 g Glass bottle 1g
10061034 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10051034 Supplier Thermo Scientific Acros Supplier No. 398170010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7304-32-7
Molecular Formula C7H4FNO4
Molecular Weight (g/mol) 185.11
MDL Number MFCD00134238
InChI Key ICXSHFWYCHJILC-UHFFFAOYSA-N
Synonym 2-fluoro-5-nitrobenzoic acid, 2-fluoro-5-nitrobenzoicacid, 2-fluoro-5-nitro-benzoic acid, benzoic acid, 2-fluoro-5-nitro, 2-fluoro-5-nitro-benzoicacid, pubchem1307, acmc-209op6, ksc377e3h, 2-fluoro-5-nitrobenzotc acid, 2-fluoro-5-nitro benzoic acid
PubChem CID 280997
IUPAC Name 2-fluoro-5-nitrobenzoic acid
SMILES OC(=O)C1=CC(=CC=C1F)[N+]([O-])=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 142.0°C to 144.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula NO2C6H3(F)COOH
Quantity 1 g
Formula Weight 185.11
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-5-nitrobenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluoro-5-nitrobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.