missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-nitrobenzotrifluoride, 98%

Product Code. 11479286
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11479286

Brand: Thermo Scientific Alfa Aesar A18130.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 400-74-8 | C7H3F4NO2 | 209.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 400-74-8
Molecular Formula C7H3F4NO2
Molecular Weight (g/mol) 209.1
MDL Number MFCD00024576
InChI Key DNTHMWUMRGOJRY-UHFFFAOYSA-N
Synonym 2-fluoro-5-nitrobenzotrifluoride, 1-fluoro-4-nitro-2-trifluoromethyl benzene, benzene, 1-fluoro-4-nitro-2-trifluoromethyl, alpha,alpha,alpha-2-tetrafluorotoluene, 2-fluoro-5-nitro trifluoromethyl benzene, 1-fluoro-4-nitro-2-trifluoromethyl-benzene, 1-fluoro-2-trifluoromethyl-4-nitrobenzene, 5-nitro-alpha,alpha,alpha,2-tetrafluorotoluene, pubchem1653, 2-fluor-5-nitrobenzotrifluorid
PubChem CID 94954
IUPAC Name 1-fluoro-4-nitro-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(F)(F)F)F

Specifications

Density 1.522
Boiling Point 105°C to 110°C (25 mmHg)
Flash Point 91°C (195°F)
Refractive Index 1.465
Quantity 5 g
UN Number UN2810
Beilstein 1881423
Formula Weight 209.1
Percent Purity 98%
Chemical Name or Material 2-Fluoro-5-nitrobenzotrifluoride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.