missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(trifluoromethoxy)aniline, 97%, Thermo Scientific™

Product Code. 15494777
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15494777 1 g 1g
15404787 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15494777 Supplier Thermo Scientific Alfa Aesar Supplier No. H32103.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
In discovery of novel benzimidazoles as potent inhibitors of TIE-2 and VEGFR-2 tyrosine kinase receptors 2-Fluoro-5-(trifluoromethoxy)aniline is used as coupling agent. 2-Fluoro-5-(trifluoromethyl)aniline may be used in chemical synthesis studies.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116369-23-4
Molecular Formula C7H5F4NO
Molecular Weight (g/mol) 195.12
MDL Number MFCD09832254
InChI Key HQUWXEXDIIWBBY-UHFFFAOYSA-N
PubChem CID 14439308
IUPAC Name 2-fluoro-5-(trifluoromethoxy)aniline
SMILES NC1=C(F)C=CC(OC(F)(F)F)=C1

Specifications

Refractive Index 1.4485
Quantity 1 g
UN Number UN2810
Synonym 2-fluoro-5-trifluoromethoxy aniline, benzenamine,2-fluoro-5-trifluoromethoxy, benzenamine, 2-fluoro-5-trifluoromethoxy, acmc-20adyv, pubchem12670, synquest 4656-3-37, 2-fluoro-3-trifluoromethoxy aniline, 2-fluoro-5-trifluoromethoxy phenylamine, 2-fluoro-5-trifluoromethoxy aniline,jrd
Solubility Information Insoluble in water.
Formula Weight 195.12
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-(trifluoromethoxy)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.