missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(trifluoromethoxy)benzyl bromide, 97%

Product Code. 11336899
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336899 1 g 1g
11346899 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11336899 Supplier Thermo Scientific Alfa Aesar Supplier No. H26795.03
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 86256-24-8 | C8H5BrF4O | 273.025 g/mol

2-Fluoro-5-(trifluoromethoxy)benzyl bromide is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-5-(trifluoromethoxy)benzyl bromide is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86256-24-8
Molecular Formula C8H5BrF4O
Molecular Weight (g/mol) 273.025
MDL Number MFCD04115890
InChI Key KOISGBFWQAPLFE-UHFFFAOYSA-N
Synonym 2-fluoro-5-trifluoromethoxy benzyl bromide, 2-bromomethyl-1-fluoro-4-trifluoromethoxy benzene, 1-bromomethyl-2-fluoro-5-trifluoromethoxy benzene
PubChem CID 2783326
IUPAC Name 2-(bromomethyl)-1-fluoro-4-(trifluoromethoxy)benzene
SMILES C1=CC(=C(C=C1OC(F)(F)F)CBr)F

Specifications

Refractive Index 1.466
Quantity 1 g
UN Number UN3265
Solubility Information Difficult to mix.
Formula Weight 273.02
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-(trifluoromethoxy)benzyl bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.