missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(trifluoromethyl)benzoic acid, 98+%

Product Code. 11439806
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11439806 1 g 1g
11449806 5 g 5g
11389619 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11439806

Brand: Thermo Scientific Alfa Aesar B20293.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 115029-23-7 | C8H3F4O2 | 207.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115029-23-7
Molecular Formula C8H3F4O2
Molecular Weight (g/mol) 207.10
MDL Number MFCD00061292
InChI Key LIFKXWNFWIUMJT-UHFFFAOYSA-M
Synonym 2-fluoro-5-trifluoromethyl benzoic acid, 2-fluoro-5-trifluoromethyl benzoicacid, alpha,alpha,alpha,6-tetrafluoro-m-toluic acid, benzoic acid, 2-fluoro-5-trifluoromethyl, maybridge1_006396, 2-fluoro-5-trifluoromethylbenzioc acid, pubchem4758, acmc-1bvsf, ksc180k3f
PubChem CID 688292
IUPAC Name 2-fluoro-5-(trifluoromethyl)benzoic acid
SMILES [O-]C(=O)C1=CC(=CC=C1F)C(F)(F)F

Specifications

Melting Point 101°C to 102°C
Quantity 1 g
Beilstein 5813056
Formula Weight 208.11
Percent Purity ≥98%
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)benzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.