missing translation for 'onlineSavingsMsg'
Learn More

2-fluoro-6-iodobenzonitrile 98%, ACROS Organics™

Product Code. 10631695 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10631695 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10631695 Supplier Thermo Scientific Acros Supplier No. 380740050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 79544-29-9
CAS Min % 97.5
CAS Max % 100.0
Melting Point 49°C to 53°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H3FIN
Linear Formula IC6H3(F)CN
MDL Number MFCD00015478
Quantity 5 g
Synonym 2-iodo-6-fluorobenzonitrile, 2-cyano-3-fluoro-1-iodobenzene, benzonitrile, 2-fluoro-6-iodo, 6-fluoro-2-iodobenzenecarbonitrile, 2-fluoro-6-iodo-benzonitrile, 6-iodo-2-fluorobenzonitrile, pubchem4780, acmc-209php, ksc494e4h, pharmabridge p-2313
InChI Key FAACTMVXBNSPJA-UHFFFAOYSA-N
SMILES FC1=CC=CC(I)=C1C#N
IUPAC Name 2-fluoro-6-iodobenzonitrile
Molecular Weight (g/mol) 247.01
PubChem CID 522722
Formula Weight 247.01
Percent Purity ≥97.5%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Fluoro-6-iodobenzonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.