missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-N-methylbenzylamine, 95%

Product Code. 15450738
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15450738 5 g 5g
15440738 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15450738 Supplier Thermo Scientific Alfa Aesar Supplier No. H54439.06
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 399-30-4 | C8H10FN | 139.173 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents and air. Store at 4-2°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 399-30-4
Molecular Formula C8H10FN
Molecular Weight (g/mol) 139.173
MDL Number MFCD04629633
InChI Key AHIHZCXUWGORQO-UHFFFAOYSA-N
Synonym n-methyl-2-fluorobenzylamine, 2-fluorobenzyl methylamine, 2-fluoro-n-methylbenzylamine, 2-fluorophenyl methyl methyl amine, 1-2-fluorophenyl-n-methylmethanamine, n-2-fluorobenzyl-n-methylamine, n-methyl-o-fluorobenzylamine, benzenemethanamine, 2-fluoro-n-methyl, 1-2-fluorophenyl-n-methyl-methanamine, acmc-209zio
PubChem CID 2060842
IUPAC Name 1-(2-fluorophenyl)-N-methylmethanamine
SMILES CNCC1=CC=CC=C1F

Specifications

Color Colorless to Tan
Density 1.047 g/mL
Boiling Point 73°C
Flash Point 68°C (154°F)
Refractive Index 1.047
Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 139.17
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Fluoro-N-methylbenzylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.