missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzonitrile, 99%

Product Code. 11446967
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
250 g
Unit Size:
100g
10g
250g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11446967 250 g 250g
11426967 10 g 10g
11436967 50 g 50g
11359619 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11446967 Supplier Thermo Scientific Alfa Aesar Supplier No. A14375.30
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 394-47-8 | C7H4FN | 121.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 394-47-8
Molecular Formula C7H4FN
Molecular Weight (g/mol) 121.11
MDL Number MFCD00001773
InChI Key GDHXJNRAJRCGMX-UHFFFAOYSA-N
Synonym o-fluorobenzonitrile, benzonitrile, 2-fluoro, o-cyanofluorobenzene, 2-fluorobenzonitril, 2-cyanofluorobenzene, benzonitrile, o-fluoro, fluorobenzonitrile, 2-fluorobenzenecarbonitrile, 2-fluoro-benzonitrile, 1-cyano-2-fluorobenzene
PubChem CID 67855
IUPAC Name 2-fluorobenzonitrile
SMILES FC1=CC=CC=C1C#N

Specifications

Density 1.138
Boiling Point 196°C to 198°C
Flash Point 73°C (163°F)
Odor Almond-like
Refractive Index 1.507
Quantity 250 g
Beilstein 2042184
Formula Weight 121.11
Percent Purity 99%
Chemical Name or Material 2-Fluorobenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.