missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzophenone, 98+%

Product Code. 11367336
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367336 5 g 5g
11377336 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367336 Supplier Thermo Scientific Alfa Aesar Supplier No. L09316.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 342-24-5 | C13H9FO | 200.212 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 342-24-5
Molecular Formula C13H9FO
Molecular Weight (g/mol) 200.212
MDL Number MFCD00000318
InChI Key DWFDQVMFSLLMPE-UHFFFAOYSA-N
Synonym 2-fluorobenzophenone, 2-fluorophenyl phenyl methanone, o-fluorobenzophenone, fluorobenzophenone, 2-fluorophenyl-phenylmethanone, 2-fluoro-phenyl-phenylmethanone, 2-fluorophenyl phenyl ketone, methanone, 2-fluorophenyl phenyl, pubchem4228, acmc-1ckt6
PubChem CID 67650
IUPAC Name (2-fluorophenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CC=C2F

Specifications

Density 1.186
Boiling Point 149°C to 150°C (16 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5864
Quantity 5 g
Beilstein 2047045
Formula Weight 200.21
Percent Purity ≥98%
Chemical Name or Material 2-Fluorobenzophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.