missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzotrifluoride, 99%

Product Code. 11419246
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419246 100 g 100g
11349619 5 g 5g
11409246 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419246 Supplier Thermo Scientific Alfa Aesar Supplier No. A18095.22
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 392-85-8 | C7H4F4 | 164.103 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 392-85-8
Molecular Formula C7H4F4
Molecular Weight (g/mol) 164.103
MDL Number MFCD00000374
InChI Key BGVGHYOIWIALFF-UHFFFAOYSA-N
Synonym 2-fluorobenzotrifluoride, 1-fluoro-2-trifluoromethyl benzene, o-fluorobenzotrifluoride, benzene, 1-fluoro-2-trifluoromethyl, alpha,alpha,alpha,2-tetrafluorotoluene, benzene, fluoro trifluoromethyl, o,alpha,alpha,alpha-tetrafluorotoluene, a,a,a,2-tetrafluorotoluene, alpha,alpha,alpha,ar-tetrafluorotoluene, 2-trifluoromethyl fluorobenzene
PubChem CID 9807
IUPAC Name 1-fluoro-2-(trifluoromethyl)benzene
SMILES C1=CC=C(C(=C1)C(F)(F)F)F

Specifications

Melting Point -49°C
Density 1.293
Boiling Point 114°C to 115°C
Flash Point 17°C (62°F)
Refractive Index 1.406
Quantity 100 g
UN Number UN1993
Beilstein 2046638
Formula Weight 164.1
Percent Purity 99%
Chemical Name or Material 2-Fluorobenzotrifluoride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Fluorobenzotrifluoride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Fluorobenzotrifluoride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.