missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzyl Chloride 98%, ACROS Organics™

Product Code. 10744421 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
100mL; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10744421 100mL; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10744421 Supplier Thermo Scientific Acros Supplier No. 119411000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 345-35-7
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2160g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H6ClF
Linear Formula FC6H4CH2Cl
Refractive Index 1.5140 to 1.5160 (20°C, 589nm)
MDL Number MFCD00000892
Quantity 100 mL
Synonym 2-fluorobenzyl chloride, 1-chloromethyl-2-fluorobenzene, o-fluorobenzyl chloride, alpha-chloro-o-fluorotoluene, benzene, 1-chloromethyl-2-fluoro, alpha-chloro-2-fluorotoluene, 2-fluorobenzylchloride, 1-chloromethyl-2-fluoro-benzene, toluene, alpha-chloro-o-fluoro, o-fluorobenzylchloride
InChI Key MOBRMRJUKNQBMY-UHFFFAOYSA-N
SMILES FC1=CC=CC=C1CCl
IUPAC Name 1-(chloromethyl)-2-fluorobenzene
Molecular Weight (g/mol) 144.57
PubChem CID 9580
Formula Weight 144.58
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 2-Fluorobenzyl chloride, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.