missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzyl chloride, 98+%

Product Code. 11404623
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11404623 25 g 25g
11414623 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11404623 Supplier Thermo Scientific Alfa Aesar Supplier No. A11566.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 345-35-7 | C7H6ClF | 144.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 345-35-7
Molecular Formula C7H6ClF
Molecular Weight (g/mol) 144.57
MDL Number MFCD00000892
InChI Key MOBRMRJUKNQBMY-UHFFFAOYSA-N
Synonym 2-fluorobenzyl chloride, 1-chloromethyl-2-fluorobenzene, o-fluorobenzyl chloride, alpha-chloro-o-fluorotoluene, benzene, 1-chloromethyl-2-fluoro, alpha-chloro-2-fluorotoluene, 2-fluorobenzylchloride, 1-chloromethyl-2-fluoro-benzene, toluene, alpha-chloro-o-fluoro, o-fluorobenzylchloride
PubChem CID 9580
IUPAC Name 1-(chloromethyl)-2-fluorobenzene
SMILES FC1=CC=CC=C1CCl

Specifications

Density 1.216
Boiling Point 175°C
Flash Point 57°C (134°F)
Odor Pungent
Refractive Index 1.5154
Quantity 25 g
UN Number UN2920
Beilstein 471699
Formula Weight 144.58
Percent Purity ≥98%
Chemical Name or Material 2-Fluorobenzyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.